Learn More
MFG Tray Nesting Storage Boxes
Nesting box
Supplier:  MFG Tray 9301085126
 View more versions of this product
Catalog No. 1523972
Only null left
Description
Description
  • Impervious to radiation, cutting oils, greases, solvents, detergents, mild acids and alkaline solutions within a pH rating of 3.0 to 10.0
  • Temperature rating of -51° to +121°C (-60° to +250°F)
  • Smooth outside surface and radial edges
  • Tough composite construction
  • Nesting design saves space
  • Machine integration and robotic applications
Specifications
Chemical Identifiers
Storage Bin
Specifications
Storage Bin
Yellow
0.2 cu. ft.
12.4 x 9.75 x 2.1 in.
11.5 in.
29.21 cm
2.125 in.
5.08 cm
Fiberglass
Rectangular
Handling and Storage
12.375 in.
29.21 cm
9.75 in.
2 in.
Product Suggestions
Product Suggestions