Electron Microscopy Sciences Block-Out Embedding Mold Release 150 ML
Catalog No. 501928627
Base mold release compound. Excellent for use as a base mold coating to allow for easy separation of paraffin from the base mold. Helps to prevent paraffin build-up on metal base molds.
Catalog No.
501928627
Supplier
Electron Microscopy SciencesSupplier No.
70899
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Base mold release compound. Excellent for use as a base mold coating to allow for easy separation of paraffin from the base mold. Helps to prevent paraffin build-up on metal base molds.