Learn More
Scientific Industries Inc MOISTURE ANALYZER 60G X .001G DFS Item
Supplier:  Scientific Industries Inc AGS60
Moisture Analyzer - Professional, 0.01% [60g x 0.001g] 250C with 250W of drying power up to 250C, heavy duty all metal housing design, precision AG electromagnetic load cell, equipped with USB, RS232, and PS2 ports which allow for quick and stable PC connection.
Catalog No. 501139358
Only null left
Explore available promotions