Learn More
Epredia™ Modified Grocott' s Methenamine Silver (Periodic Acid) Stain Kit
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid)
$408.50 - $1217.10
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|
23-900-656
|
Epredia™
SH87008 |
Silver Stain Kit, Periodic Acid |
Each for $1,217.10
|
|
|||||
|
22-110-652
|
Epredia™
88026 |
Gold Chloride Solution (0.1%) |
Each for $408.50
|
|
|||||
Description
- Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
- Background is stained green