Learn More
Model Plus The Atom
Catalog No. S94391
Change view
Click to view available options
Grade:
6 to 10
Catalog No. Grade
S94391 6 to 10
1 options
Catalog No. S94391 Supplier Customer Channels Group (Ccg) Supplier No. 69077
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Illustrates latest scientific discovery to students.
  • Shows electron orbits as having the appearance of a fuzzy electron cloud
  • Grades 6 to 10
Specifications
Grade 6 to 10
Includes Shows electron orbits as having the appearance of a fuzzy electron cloud
Product Type Atom Model