missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bullard™ 88VX Series Airline Respirator Accessories, Nylon Capes
Tan nylon replacement capes for use with 88VX supplied-air respirator systems.
$415.68 - $415.68
Specifications
| Color | Tan |
|---|---|
| For Use With (Equipment) | 88VX Series Respirator |
| Material | Nylon |
| Includes | Snap Hooks, Breathing Tube Strap, Additional Nylon Liner in Neck Cuff Area |
| Product Type | Cape |
Specifications
| Tan | |
| Nylon | |
| Cape |
| 88VX Series Respirator | |
| Snap Hooks, Breathing Tube Strap, Additional Nylon Liner in Neck Cuff Area |