Learn More
Model 400 Portable Fume Extractor
Catalog No. NC1670998
Encompass
The Portable Fume Extractor provides a powerful engineering safety control ideal for controlling fumes and particulate in a medical or laboratory environment. This system uses an adjustable 72 in. (182.88 cm) long self-supportive arm to capture particulate and chemical fumes at the source and draw them into the filtration system. Then, the filtered air is released back into the surrounding room creating a recirculating airflow pattern. The Portable Fume Extractor offers high portability with a handle and heavy-duty casters to easily allow movement to wherever fume extraction is needed. This system has an easy-to-change filter and variable speed control as standard features. Multiple types of filters can be used at once to capture both fumes and particulate.
Catalog No. NC1670998
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Portable Fume Extractor provides a powerful engineering safety control ideal for controlling fumes and particulate in a medical or laboratory environment. This system uses an adjustable 72 in. (182.88 cm) long self-supportive arm to capture particulate and chemical fumes at the source and draw them into the filtration system. Then, the filtered air is released back into the surrounding room creating a recirculating airflow pattern. The Portable Fume Extractor offers high portability with a handle and heavy-duty casters to easily allow movement to wherever fume extraction is needed. This system has an easy-to-change filter and variable speed control as standard features. Multiple types of filters can be used at once to capture both fumes and particulate.