Learn More
Emerson Apparatus MODEL 145 SPEED DRYER
Numéro de catalogue. NC0601839
Encompass
  • Control accuracy of 1 C at 105 C
  • Temperature range up to 150 C (230 C available on request)
  • Plate area of 14 in x 24 in
  • Surface temperature linearity of +/- 2.5 C (entire plate) and <+/- 0.1 C/cm (except perimeter)
  • Input voltage options of 110 VAC or 220 VAC
  • Power of 1000 Watts (primary), 1500 Watts (secondary)
  • Weight of 45 lbs
  • Shipping dimensions of 30 in x 9 in x 20 in
  • Non-conductive cover handle for user safety
  • Multilayer ceramic insulation
  • Dielectric distributed heat plate
  • Digital set point controller and heavy duty solid state relay
  • Supports TAPPI, CPPA, ASTM, APPITA test methods
Numéro de catalogue. NC0601839 Fournisseur Emerson Apparatus Code fournisseur 032504
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
  • Control accuracy of 1 C at 105 C
  • Temperature range up to 150 C (230 C available on request)
  • Plate area of 14 in x 24 in
  • Surface temperature linearity of +/- 2.5 C (entire plate) and <+/- 0.1 C/cm (except perimeter)
  • Input voltage options of 110 VAC or 220 VAC
  • Power of 1000 Watts (primary), 1500 Watts (secondary)
  • Weight of 45 lbs
  • Shipping dimensions of 30 in x 9 in x 20 in
  • Non-conductive cover handle for user safety
  • Multilayer ceramic insulation
  • Dielectric distributed heat plate
  • Digital set point controller and heavy duty solid state relay
  • Supports TAPPI, CPPA, ASTM, APPITA test methods