missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mMESSAGE mMACHINE™ T3 Transcription Kit

Catalog No. AM1348
Encompass
Change view
Click to view available options
Product Type:
T3 In Vitro Transcription kit
No. of Reactions:
25 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type No. of Reactions
AM1348 T3 In Vitro Transcription kit 25 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1348 Supplier Invitrogen™ Supplier No. AM1348

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Obtain 10–50 times yield of conventional in vitro transcription reactions.

mMESSAGE mMACHINE high yield capped RNA transcription kits use patented high-yield transcription technology for the routine synthesis of 15–35 μg of 7-methyl guanosine capped RNA from 1 μg of template DNA. Each kit includes sufficient reagents for 25 reactions.

  • High yields of capped RNA—up to 35 μg of RNA per reaction
  • Extra convenience—cap analog-nucleotide mix provided
  • Simplified reaction format
  • Synthesized transcripts ideal for microinjection, in vitro translation, or transfection
  • 10–50 times yield obtained with conventional in vitro transcription reactions
  • Ratio of cap analog to GTP has been optimized to allow compromise between yield (15–35 μg) and proportion of transcripts that are capped (80%)
  • Contains a LiCl precipitation solution, efficient for separating proteins and unincorporated nucleotides (including free cap analog) from the capped RNA, allowing an increased efficiency of translation
  • Optimized for use with polymerases included in kit
  • Contains 57 U of cap analog

Specifications

Content And Storage T3 Enzyme Mix, 10X Reaction Buffer 2XNTP/CAP Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, and Gel Loading Buffer II are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Final Product Type Synthetic mRNA, mRNA
Format Kit
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions 25 Reactions
Product Line Ambion, mMESSAGE mMACHINE
Product Type T3 In Vitro Transcription kit
Quantity 25 Reactions
Promoter T3
Shipping Condition Dry Ice
Yield 20-30 μg
Starting Material PCR Products, Linearized DNA plasmid
Show More Show Less
I am using the mMESSAGE mMACHINE T3 Transcription Kit for in vitro transcription. I ran my reaction product on a denaturing gel and got a smear. Can you offer some advice?

If the RNA appears degraded (e.g. smeared), remove residual RNase from the DNA template preparation before in vitro transcription. Do this by digesting the DNA prep with proteinase K (100-200 µg/mL) in the presence of 0.5% SDS for30 min at 50°C, follow this with phenol/chloroform extraction. The RNase Inhibitor that is present in the transcription reaction, can only inactivate trace RNase contamination. Large amounts of RNase contamination will compromise the size and amount of transcription products.

For Research Use Only. Not for use in diagnostic procedures.