missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Mixtainer BioProcess Container (BPC) for HyPerforma™ Mixtainer™ Systems

Description
Recommended for:
- Hydration of powdered media and buffers
- Sterile mixing of cell culture media with sera and reagents
- Sterile mixing of buffer solutions or other process fluids
- Pooling of sterile liquid fractions
Specifications
Specifications
| Capacity (Metric) | 50 L |
| Port Detail | 4 x Top-mounted Ports and Tubing |
| Tubing Line 1 | 0.25 in. ID Tubing and male/female Luer connectors (Port 1) |
| Tubing Line 2 | 0.25 in. ID Tubing and male/female Luer connectors (Port 2) |
| Tubing Line 3 | 3/8 in. ID tubing and quick-connect couplings (Port 3) |
| Tubing Line 4 | 3/8 in. ID tubing and quick-connect couplings (Port 4) |
| Tubing Style | TPE |
| Sterility | Sterile |