Learn More
Thermo Scientific™ Mixtainer BioProcess Container (BPC) for HyPerforma™ Mixtainer™ Systems
GREENER_CHOICE
Catalog No. SH3068701
Change view
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
Port Detail:
4 x Top-mounted Ports and Tubing
Powder Port
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Port Detail
SH3068701 50 L Powder Port
SH3066101 50 L 4 x Top-mounted Ports and Tubing
SH3066102 100 L 4 x Top-mounted Ports and Tubing
SH3066103 200 L 4 x Top-mounted Ports and Tubing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SH3068701 Supplier Thermo Scientific™ Supplier No. SH30687.01
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The complete Thermo Scientific™ Mixtainer™ System is an integrated, single-use sterile unit for mixing of cell culture media, and other process liquids such as buffers, reagents and bulk drug precursors or products.
Recommended for:
  • Hydration of powdered media and buffers
  • Sterile mixing of cell culture media with sera and reagents
  • Sterile mixing of buffer solutions or other process fluids
  • Pooling of sterile liquid fractions
Specifications
Capacity (Metric) 50 L
Port Detail Powder Port
Tubing Style TPE