Learn More
Thermo Scientific™ Mixtainer BioProcess Container (BPC) for HyPerforma™ Mixtainer™ Systems
The complete Thermo Scientific™ Mixtainer™ System is an integrated, single-use sterile unit for mixing of cell culture media, and other process liquids such as buffers, reagents and bulk drug precursors or products.
Supplier: Thermo Scientific™ SH30661.02
Description
Recommended for:- Hydration of powdered media and buffers
- Sterile mixing of cell culture media with sera and reagents
- Sterile mixing of buffer solutions or other process fluids
- Pooling of sterile liquid fractions
Identifiants chimiques
| TPE |
Spécifications
| 100 L | |
| 0.25 in. ID Tubing and male/female Luer connectors (Port 1) | |
| 3/8 in. ID tubing and quick-connect couplings (Port 3) | |
| TPE |
| 4 x Top-mounted Ports and Tubing | |
| 0.25 in. ID Tubing and male/female Luer connectors (Port 2) | |
| 3/8 in. ID tubing and quick-connect couplings (Port 4) | |
| Sterile |