missing translation for 'onlineSavingsMsg'
Learn More

Mixed Acid, 10% (v/v) Hydrofluoric Acid/10% (v/v) Nitric Acid, Ricca Chemical

Catalog No. r51513001a
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R51513001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R51513001A Supplier Ricca Chemical Company Supplier No. R51513001A
Only null left

Specifications

Chemical Name or Material 10% (v/v) Nitric acid-10% (v/v) hydrofluoric acid mixtures
Packaging Natural Poly Bottle
CAS 7664-39-3, 7697-37-2, 7732-18-5
Physical Form Liquid
Color Colorless
pH < 2
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Solution
Type Analytical Volumetric Solution
Quantity 1 L
Percent Purity 0.1
Grade Laboratory
Show More Show Less