missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mitoxantrone Dihydrochloride

Catalog No. AAJ66341MF
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66341MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66341MF Supplier Thermo Scientific Chemicals Supplier No. J66341MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70476-82-3 | C22H30Cl2N4O6, C22H28N4O6.2HCl | 517.40 g/mol

Mitoxantrone dihydrochloride is an antiviral, antibacterial, antiprotozoal, immunomodulating, and antineoplastic cytostatic anthraquinone derivative. Induces DNA damage by intercalating into DNA and inhibiting Topo II (topoisomerase II). Mitoxantrone dihydrochloride induces interstrand DNA cross-links and DNA-protein cross-links in cellular systems. Mitoxantrone dihydrochloride has recently been shown to be an inhibitor of DNA methylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70476-82-3
Molecular Formula C22H28N4O6.2HCl
Synonym 1,4-Dihydroxy-5,8-bis[[2-[(2-hydroxyethyl)amino]ethyl]amino]-9,10-anthracenedione dihydrochloride;

Specifications

Quantity 50 mg
Description Blue to black
Content And Storage -30°C to -10°C
Formula Weight 517.40
Solubility Soluble to 5 mM in water and to 75 mM in DMSO
Product Type Mitoxantrone dihydrochloride
Form Crystalline solid

RUO – Research Use Only