Learn More
Invitrogen™ MitoProbe™ Transition Pore Assay Kit, for flow cytometry
Description
View a selection guide for all apoptosis assays for flow cytometry.
Specifications
Specifications
| Quantity | 1 kit |
| Format | Tube |
| Product Type | Transition Pore Assay Kit |
| Content And Storage | Contains 5 vials of calcein, AM (50 μg of dry powder/vial), 1 vial of cobalt (II) Chloride hexahydrate (1.2 mL, 80 mM in saline), 1 vial of ionomycin (55 μg), calcium salt, and 1 vial of DMSO (1.5 mL). Store in freezer (-5 to -30°C). |
| Excitation/Emission | 488 |
| No. of Reactions | 100 Reactions |
| Shipping Condition | Room Temperature |
| Product Line | MitoProbe |
| Conjugate | Calcein AM |
Frequently Asked Questions (FAQs)
-Measures data from single cells.
-Data are obtained for a large number of cells, generating a rich statistical analysis of cell populations.
-Because single cells are measured, it will reveal heterogeneity within a population.
-With the ability to multiplex, small sub-populations can be identified.
-Thousands of cells can be analyzed rapidly.
-It is ideally suited for blood samples and other cells in suspension.
-Data can be re-analyzed multiple times after acquisition.
-Flow cytometry files (FCS) can be archived.
There are several applications, some of which include immunophenotyping, cell cycle analysis, apoptosis assays such as annexin V staining, CellEvent Caspase-3/7 assay, and TUNEL assay, cell viability, proliferation assays such as CellTrace assay and Click-iT EdU assay, measurements of mitochondrial potential with MitoProbe assays, and cell counting using counting beads.
For Research Use Only. Not for use in diagnostic procedures.