Learn More
Selleck Chemical Llc Mitomycin C 10mg 50-07-7 DFS Item
Supplier:  Selleck Chemicals Co Ltd S814610MG
Mitomycin C is an antineoplastic antibiotic by inhibiting DNA synthesis, used to treat different cancers. Mitomycin C induces apoptosis in a caspases-dependent and Fas/CD95-independent manner. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
Catalog No. NC1081159
Only null left
Explore available promotions