missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mitocondrial Translational Initiation Factor 3 Recombinant Protein Antigen
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human MTIF3. The Mitocondrial Translational Initiation Factor 3 Recombinant Protein Antigen is derived from E. coli. The Mitocondrial Translational Initiation Factor 3 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
This is a blocking peptide for NBP1-92121. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.
Specifications
Specifications
| Gene ID (Entrez) | 219402 |
| Species | Human |
| Product Line | Novus |
| Purification Method | Chromatography |
| Purity | >80% |
| Concentration | 0.5mg/mL |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | Blocking/Neutralizing, Control |
| Gene Symbol | MTIF3 |
| Show More |
For Research Use Only