Learn More
Abcam Inc Mithramycin A (Plicamycin), DNA and RNA polymerase inhibitor, 1MG
Catalog No. 502706199
Encompass
MW 1085.1 Da, Purity >98%. DNA and RNA polymerase inhibitor, DNA binding transcriptional inhibitor (Sp1 and Sp3). Shows antineoplastic activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502706199 Supplier Abcam Inc Supplier No. AB1427231MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 1085.1 Da, Purity >98%. DNA and RNA polymerase inhibitor, DNA binding transcriptional inhibitor (Sp1 and Sp3). Shows antineoplastic activity.
The product is subject to the following: Abcam Restricted Use Statement