missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Misoprostol, free acid, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 mg |
| UN Number | UN2810 |
| Formula Weight | 368.51 |
| Physical Form | Liquid |
| Chemical Name or Material | Misoprostol, Free acid |
Safety and Handling
| Hazard Category | H301-H315-H319-H335-H360 |
| Hazard Statement | GHS H Statement H301-H360-H315-H319-H335 Toxic if swallowed. May damage fertility or the unborn child. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
| TSCA | No |
| storageNote1 | -30° to -10°C |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only