Learn More
Mirus Bio™ Ingenio™ Cell Droppers for Electroporation
Catalog No. MIR50125
Change view
Click to view available options
Quantity:
50 Pk.
Catalog No. Quantity
MIR50125 50 Pk.
1 options
Catalog No. MIR50125 Supplier Mirus Bio™ Supplier No. MIR 50125
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ingenio™ Cell Droppers for Electroporation for use with the Ingenio Electroporation System.
  • Deliver to hard to transfect cell lines and primary cells
  • Use your existing system including Ingenio EZporator, Lonza-Amaxa, Bio-Rad or Harvard BTX
  • Ingenio Electroporation Solution is available as a stand-alone solution or as part of a complete kit with cuvettes and cell droppers
Specifications
Sterility Sterile
Type Cell Dropper
Disposable Single-use
Material Polycarbonate
Includes 50 Cell Droppers
Quantity 50 Pk.
Width (Metric) Gap 0.2 cm
For Use With (Equipment) Ingenio Electroporation Kit
For Use With (Application) Electroporation