missing translation for 'onlineSavingsMsg'
Learn More

Mirtazapine 98.0+%, TCI America™

Catalog No. p-7133054
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2151100MG 100 mg
M21511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2151100MG Supplier TCI America Supplier No. M2151100MG
Only null left

Chemical Identifiers

CAS 85650-52-8
Molecular Formula C17H19N3
Molecular Weight (g/mol) 265.36
MDL Number MFCD00865427
InChI Key RONZAEMNMFQXRA-UHFFFAOYSA-N
Synonym mirtazapine, remeron, mepirzepine, remergil, zispin, remergon, rexer, 6-azamianserin, remeron soltab, mepirzapin
PubChem CID 4205
ChEBI CHEBI:6950
SMILES CN1CCN2C(C1)C3=CC=CC=C3CC4=C2N=CC=C4

Specifications

Melting Point 121°C
Color White-Yellow
Quantity 100 mg
Formula Weight 265.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Mirtazapine
RTECSNumber UQ4410250
TSCA No