Learn More
Minuteman™ Tool Kit for Wet/Dry HEPA Vacuum Systems
Catalog No. 19811787
Encompass
Change view
Click to view available options
Product Type:
Dry Vacuum Tool Kit
Wet/Dry Vacuum Tool Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19811787 Dry Vacuum Tool Kit
18999983 Wet/Dry Vacuum Tool Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19811787 Supplier Minuteman™ Supplier No. 4900221
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
0.5 in. × 10 ft. (1.3 x 25.4cm) crushproof hose, 1.5 in. (3.8cm) wand, 12 in. (30.5cm) crevice tool, round dust brush, 5 in. (12.7cm) upholstery tool, 5 in. (12.7cm)slide-on bristle brush, brush tool, 11 in. (28cm) carpet/floor tool, and squeegee tool assembly.
For use with 4- and 6-gallon vacuums (with plastic intake) and 15- and 20-gallon vacuums.
Specifications
Includes 0.5 in. x 10 ft. Crushproof Hose, 1.5 in. Wand, 12 in. Crevice Tool, 5 in. Upholstery Tool, 5 in. Slide-on Bristle Brush, Brush Tool, 11 in. Carpet or floor Tool
Product Type Dry Vacuum Tool Kit
For Use With (Equipment) Any 290 Dry only, 390 Dry only, any Critical Filter Dry only