missing translation for 'onlineSavingsMsg'
Learn More

Minuteman™ Flammable Liquid Recovery Vacuum

Catalog No. p-3650589
Encompass
Change view
Click to view available options
Product Type:
Flammable Liquid Vacuum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
18063502 Flammable Liquid Vacuum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18063502 Supplier Minuteman™ Supplier No. C87355 01

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Special neoprene 1.5 in. x 25 ft. (3.8cm x 7.6m) grounded vacuum hose with nitrile core; explosion-proof operator handle; all steel dolly with 5 in. dia. 912.7cm) conductive wheels for easy transport; and an explosion-proof tool kit containing an explosion-proof hose, operator's handle/wand with a 14 in. (35.6cm) floor squeegee tool, and a crevice tool.

Requires

Grounded air line hose assembly

For safe recovery of jet fuel and hazardous liquids

  • Ideal for use in areas where electricity is not available
  • Compressed-air operated with no moving parts
  • Provides high air flow and high static lift for recovering large amounts of liquid debris quickly and safely
  • Attached silencer for quieter operation of the system
  • Features lid assembly with locking lid latches, pressure relief valve, intake hose closure, and liquid shut-off for wet applications

Order Info

Wet capacity: 45 gal. (170L). Static lift: 180 in. (457.2cm). Air flow: 182cfm (5.1m). Input pressure: 100psi (7bar). Input air: 46cfm (1.3m/min.). Weight: 181 lb. (82kg). Air supply hose assembly with antistatic line available separately. Call your Fisher Safety Customer Service Representative for additional hose sizes.

Compliance

Meets GSA Specification for Pneumatic Vacuum Cleaner, Air-Operated for recovery of flammable liquids. National Stock No. 7910-632-9840.

Specifications

Weight (Metric) 82 kg
Includes Special Neoprene 1.5 in. x 25 ft. Grounded Vacuum Hose with Nitrile Core, Explosion Proof Operator Handle, all Steel Dolly with 5 in. dia. Conductive Wheels for Easy Transport and an Explosion Proof Tool Kit containing an Explosion Proof Hose, Operator's
Weight (English) 181 lb.
Product Type Flammable Liquid Vacuum
Capacity (English) 5760 oz.
Capacity (Metric) 170 L
Airflow Rate 182 cfm
Capacity (English) Tank 7040 oz.
Capacity (Metric) Tank 208 L
Water Lift 180 in.