Learn More
Minuteman™ Critical Filter Vacuums
Rugged, high-performance polyethylene wet/dry and dry vacuums recover asbestos and other hazardous materials
Supplier: Minuteman™ C82906 01
Includes:
Wet/Dry Vacuum supplied with polyethylene Uni-Adapter™ attachment for leakproof pickup in wet applications.
Description
- Feature multifilter systems that include ULPA filter that is 99.99% effective at 0.12μm
- Aluminum and steel body and flange make filter highly resistant to punctures, tears, abrasions and damage from moisture
- Polyethylene tank with polypropylene lid assembly to minimize noise
- Direct load to 6mil lined tank increases cleaning capacity, eliminates double handling of debris and reduces decontamination time
- Patented self-sealing intake fits 1.25, 1.5 and 2 in. hoses (3.18, 3.81 and 5.1cm); quick-release mechanism allows easy hose connection and removal
- Patented safety interlock system prevents operation if filter is misaligned
- Compact 6 gal. (23L) tank is ideal for tight areas, glove bagging, or use with AHERA kits
- Larger 15-gal. (57L) tank uses dimpled cloth bag that provides 50% more filtration than standard bags
Order Info
For 115V. Optional accessory kit includes crushproof hose; wand; round dust brush; slide-on bristle brush; and crevice, upholstery brush and squeegee floor assembly tools (order separately; for vacuum 18-999-984, order kit 18-999-983; for 18-999-986, order kit 18-999-287). Replacement ULPA filter 18-999-989 also available.
Compliance
Meet or exceed MIL-F-51079D, MIL-F-51068F, UL Standard 586 ULPA Filter Efficiency, A.E.C. Regulatory Guide 1.51, UL Class II Flammability Rating and ANSI™ Z9.2.
Specifications
| 115 V | |
| Wet or Dry Vacuum Supplied with Polyethylene Uni-Adapter™ Attachment for Leakproof Pickup in Wet Applications | |
| Abatement Contractors, First Responders, Municipalities, Primary/Higher Education, Environmental Contractors, Military, Nuclear, Ship Building, Stainless Steel Manufacturing, Welding | |
| 0.46 cu. ft. | |
| 95 cfm | |
| 22 L | |
| 26 in. | |
| 70 dB | |
| 41 cm | |
| MIL-F-51079D, MIL-F-51068F, UL, A.E.C., UL Class II, ANSI Z9.2 |
| 12 kg | |
| 27 lb. | |
| Critical Filter Vacuum | |
| 0.01 m3/min. | |
| 768 oz. | |
| ULPA | |
| 66 cm | |
| 16 in. | |
| 85 in. | |
| 50 to 60 Hz |