Learn More
Research Products Corporation Mini-Strip Blotting Box, 6 Lanes, 10.0 x 3.0 x 2.5cm, Clear, 2 per Package
Catalog No. 50550311
Encompass
Perfect size container for incubating or staining western blot strips. Clear plastic box has individual chambers that allow processing multiple strips at one time. Flat bottom on container reduces the amount of solution necessary to cover each strip. Box also features a tight fitting hinged lid that prevents evaporation. Made of high clarity polystyrene for enhanced visibility of contents.
Catalog No. 50550311 Supplier Research Products Corporation Supplier No. 248778
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Perfect size container for incubating or staining western blot strips. Clear plastic box has individual chambers that allow processing multiple strips at one time. Flat bottom on container reduces the amount of solution necessary to cover each strip. Box also features a tight fitting hinged lid that prevents evaporation. Made of high clarity polystyrene for enhanced visibility of contents.