missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Minimum Essential Medium Eagle, Liquid

Catalog No. p-4474843
Change view
Click to view available options
With Additives:
Earle's Balanced Salt Solution (EBSS), Sodium Bicarbonate
Sodium Bicarbonate, Earle's Balanced Salt Solution (EBSS)
Without Additives:
No Glutamine
No Glutamine, No Cysteine, No Cystine, No Methionine
Product Type:
Minimum Essential Medium Eagle's
Minimum Essential Medium Eagle's (modified)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. With Additives Without Additives Product Type
MP091210254 Earle's Balanced Salt Solution (EBSS), Sodium Bicarbonate No Glutamine Minimum Essential Medium Eagle's
ICN1641454 Sodium Bicarbonate, Earle's Balanced Salt Solution (EBSS) No Glutamine, No Cysteine, No Cystine, No Methionine Minimum Essential Medium Eagle's (modified)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP091210254 Supplier MP Biomedicals™ Supplier No. 091210254
Only null left

Incorporates higher amino acid concentrations for the cultivation of fastidious cells

MEM, originally prepared by Harry Eagle, is one of the most popular cell culture media. Upon his attempts to cultivate normal mammalian fibroblasts and certain HeLa cell subtypes, it was revealed that the nutritional needs of these cell types could not be met by BME. Further studies led to the development of MEM from MP Biomedicals incorporating specific modifications such as higher amino acid concentrations for the cultivation of fastidious cells.

  • MEM from MP Biomedicals may be used to support the growth of cells in monolayers, in suspension and wide variety of other cell types with proper supplementation.
  • Available with either Earle's or Hank's salts.

Specifications

Product Type Minimum Essential Medium Eagle's
Sterility Sterile
With Additives Earle's Balanced Salt Solution (EBSS), Sodium Bicarbonate
Without Additives No Glutamine
Quantity 500 mL
pH 6.9 to 7.3
Content And Storage 0 to 5°C

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.