missing translation for 'onlineSavingsMsg'
Learn More

Alcor™ miniiWASH™ Wash Fluid

Catalog No. 22000453
Encompass
Change view
Click to view available options
Quantity:
250 mL/Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22000453 250 mL/Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22000453 Supplier Alcor™ Supplier No. 11212003

missing translation for 'validMainframeMsg'

Wash fluid bottle provides enough fluid for up to 80 wash cycles.

  • Pre-filled bottle
  • With screw cap
  • 250 mL bottle

Specifications

Content And Storage Room Temperature
For Use With (Equipment) miniiSED™ automted ESR analyzer
Sample Type Whole Blood
Packaging 4/Pk
Quantity 250 mL/Bottle
Stability Stable until product expiration