missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Minigrip™ Reloc ZIPPIT™ Clear Non-Biohazard Reclosable Zipper Bags
Description
Economical bags with zipper closure
- Clear (plain) bags with no biohazard logo
- Wide range of sizes and gauges
- With or without white block and hangholes
- DispenserBag™ convenience packaging
To avoid the risk of cross-contamination, specimen bags should not be reused.
Specifications
Specifications
| Closure Type | Zip Seal |
| Color | Clear |
| Thickness (English) | 4 mil |
| Dimensions (L x W) | 15 x 12 in. (38.1 x 30.5cm) |
| Length (English) | 15 in. |
| Width (English) | 12 in. |
| Product Type | Specimen Transport Bag |
| Length (Metric) | 38.1 cm |
| Width (Metric) | 30.5 cm |