Learn More
Minigrip™ GreenLine™ Biodegradable Reclosable Zipper Bags
For use with all kinds of lab, medical and food storage
Supplier:  Minigrip™ MGBD2P0912
 View more versions of this product
Catalog No. 22306414
Only null left
Description
Description
  • Made of low-density polyethylene
  • Thickness of 2 mil
  • Bags tested to biodegrade 15% within 12 months per ASTM D5511 test results when in contact with other materials that were also biodegrading
  • Available in variety of sizes
Compliance
Proven: ASTM D5511 tested by an independent lab to be biodegradable
Specifications
Chemical Identifiers
2 mil
Specifications
2 mil
22.8 x 30.4cm (9 x 12 in.)
Product Suggestions
Product Suggestions