Learn More
Minigrip™ GreenLine™ Biodegradable Reclosable Zipper Bags
For use with all kinds of lab, medical and food storage
Supplier: Minigrip™ MGBD2P0406
Description
- Made of low-density polyethylene
- Thickness of 2 mil
- Bags tested to biodegrade 15% within 12 months per ASTM D5511 test results when in contact with other materials that were also biodegrading
- Available in variety of sizes
Compliance
Proven: ASTM D5511 tested by an independent lab to be biodegradable
Chemical Identifiers
| 2 mil |
Specifications
| 2 mil | |
| 10.16 x 15.2cm (4 x 6 in.) |