Learn More
Research Products Corporation Mini-Blotting Containers, 11.5 x 8.9 x 2.2cm, Clear, 6 per Case
Catalog No. 50550302
Encompass
Plastic containers for many laboratory applications. Containers have a smooth, flat surface that is ideal for blotting membranes or staining gels. Hinged lid with snap lock closure prevents evaporation of reagents. Constructed of ultra-clear polystyrene.
Catalog No. 50550302 Supplier Research Products Corporation Supplier No. 248717
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Plastic containers for many laboratory applications. Containers have a smooth, flat surface that is ideal for blotting membranes or staining gels. Hinged lid with snap lock closure prevents evaporation of reagents. Constructed of ultra-clear polystyrene.