missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Miniature Human Skull Model, 2.5 in.

Catalog No. S15012
Change view
Click to view available options
Dimensions (L x W x H):
2 x 1.5 x 2.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S15012 2 x 1.5 x 2.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15012 Supplier Eisco™ Supplier No. AMCH1063AS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Miniature Human Skull Model is of medical quality and provides an incredible amount of realism and detail for examining the human skull.

  • Three part skull model includes Calvaria, base of skull, and mandible
  • Mandible is articulated for more in depth study, and to demonstrate mandible movement
  • Cast and scaled down from a real human skull
  • Features realistic details, textures, and bony landmarks providing an incredible amount of realism
  • Made of durable and sturdy plastisol polymer
  • Model is easy to clean and will hold up to years of handling
  • Perfect size for individual use and virtual lessons

Specifications

Color White
Dimensions (L x W x H) 2 x 1.5 x 2.5 in.
For Use With (Application) In Depth Examination of the Human Skull
Material Plastisol Polymer
Product Type Model, Skull