Learn More
Pyramex™ Mini Ztek™ Safety Eyewear
Description
Specifications
Specifications
| Lens Coating | Anti-fog |
| Certifications/Compliance | ANSI Z87.1, CSA Z94.3-07 |
| Lens Tint | Clear |
| Weight (Metric) | 20 g |
| Lens Material | Polycarbonate |