Learn More
Mopec Mini Section Dryer
Description
- Fan-assisted heating dramatically reduces drying time
- Designed to take up minimal bench-top space
- Digital temperature control
- Rapid heating system ensures system is ready to use in less time
- Anti-microbial coating to help prevent bacteria and virus growth
- Timer Unit allows monitoring for up to four slides and ensures each rack has the correct amount of time in the dryer
- Rack Holder holds 4 slide racks and works in conjunction with the timer unit
- Dako Omnis Rack Holder holds up to nine Dako Omnis slide racks
Specifications
Specifications
| Product Type | Microscope Slide Dryer |
| For Use With (Application) | Drying Specimens |
| For Use With (Equipment) | Microtome |