Learn More
Research Products Corporation Mini Incubator, Ambient +5C to 60C, 11 x 11 x 13 Inches
Catalog No. 50476517
Encompass
Ideal compact size incubator offers an efficient and economical method for incubating culture flasks, petri dishes and plates. Features a broad temperature range to accommodate a variety of applications. Wrap around heating element provides uniform heating and is accurately maintained with a hydraulic thermostat.
Additional features include: Pilot light heating indicator; Acrylic viewing area in door; Adjustable shelf and Thermometer port.
Catalog No. 50476517 Supplier Research Products Corporation Supplier No. RPI 246265
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ideal compact size incubator offers an efficient and economical method for incubating culture flasks, petri dishes and plates. Features a broad temperature range to accommodate a variety of applications. Wrap around heating element provides uniform heating and is accurately maintained with a hydraulic thermostat.
Additional features include: Pilot light heating indicator; Acrylic viewing area in door; Adjustable shelf and Thermometer port.