missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mini Capillary Unions

Catalog No. 03050244
Change view
Click to view available options
For Use With (Equipment):
0.22 to 0.32 mm ID Capillary GC Column
0.53 mm ID Capillary GC Column
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
03050244 0.22 to 0.32 mm ID Capillary GC Column
03050245 0.53 mm ID Capillary GC Column
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03050244 Supplier Thermo Scientific™ Supplier No. 290GU498

Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.

Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.

Kit contains one union and five ferrules

Includes:

Union, 1; Ferrules, 5

Specifications

Type Capillary Union
Description For 0.32 mm I.D. Capillary
For Use With (Equipment) 0.22 to 0.32 mm ID Capillary GC Column