missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPEX CertiPrep™ Minerals Testing Kit
Description
Claritas PPT Grade Minerals Testing Kit for ICP-MS, 1,000μg/mL (1,000 ppm), 6 each 30mL
Kit contains:
- Calcium, 1,000μg/mL, for ICP-MS, 30mL, Matrix: 2% HNO3
- Iron, 1,000μg/mL, for ICP-MS, 30mL, Matrix: 2% HNO3
- Potassium, 1,000μg/mL, for ICP-MS, 30mL, Matrix: 2% HNO3
- Magnesium, 1,000μg/mL, for ICP-MS, 30mL Matrix: 2% HNO3
- Sodium, 1,000μg/mL, for ICP-MS, 30mL Matrix: 2% HNO3
Specifications
Specifications
| Description | 2% HNO3 |
| Volume (Metric) | 30 mL |
| For Use With (Equipment) | Mineral teesting |
| Quantity | 30 mL |
| Storage Requirements | Ambient |
| Type | Certified Reference Material |