missing translation for 'onlineSavingsMsg'
Learn More

Mineral Spirits, Spectrum™ Chemical

Catalog No. p-7158580
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Steel Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604630 500 mL Steel Can
18604631 4 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18604630 Supplier Spectrum Chemical Mfg Cor Supplier No. M1292500MLGL
Only null left

M1292, 64475-85-0

Spectrum™ Chemical Mineral Spirits , also known as white spirit or mineral turpentine, is a petroleum-derived liquid used as an organic solvent in decorating and painting. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 64475-85-0

Specifications

CAS Max % 1
Boiling Point 149°C to 213°C
Quantity 500 mL
Packaging Steel Can
UN Number UN1268
Grade Reagent
Chemical Name or Material Mineral Spirits