missing translation for 'onlineSavingsMsg'
Learn More

Mineral Oil, Medium, Reagents

Catalog No. p-200021093
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337170 1 L
01337168 4 L
01337167 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337170 Supplier Labchem Inc Supplier No. C22517001C
Only null left

Mineral Oil, Medium, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 8012-95-1
Molecular Formula MFCD00131611
Molecular Weight (g/mol) 452.363

Specifications

Boiling Range >230°C
Color Colorless
Boiling Point >230°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 0.85
Grade Reagent
Physical Form Liquid
Chemical Name or Material Mineral Oil, Medium