missing translation for 'onlineSavingsMsg'
Learn More

Mineral Oil, Heavy, USP, Spectrum™ Chemical

Catalog No. 18604607
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18604607 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18604607 Supplier Spectrum Chemical Mfg Cor Supplier No. MI500500MLGL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MI500, 8012-95-1

Spectrum™ Chemical Mineral Oil, Heavy, USP is a lubricant laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8012-95-1
Molecular Formula MFCD00131611
Molecular Weight (g/mol) 0.00
MDL Number MFCD00131611
InChI Key FFNDMZIBVDSQFI-UHFFFAOYSA-N
IUPAC Name 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-1λ4-chromen-1-ylium chloride
SMILES *

Specifications

Acidity To pass test
CAS Max % 1%
Boiling Point >215°C
Packaging Amber Glass Bottle
Quantity 500 mL
Viscosity 34.5-150 mm2
Grade USP
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Mineral Oil, Heavy