missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mineral Oil, Heavy, USP, Spectrum™ Chemical
MI500, 8012-95-1
Supplier: Spectrum Chemical Mfg Cor MI50020LTPL
Description
Spectrum™ Chemical Mineral Oil, Heavy, USP is a lubricant laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| To pass test | |
| 1 | |
| MFCD00131611 | |
| 20 L | |
| * | |
| 0.00 | |
| USP |
| 8012-95-1 | |
| Poly Drum | |
| MFCD00131611 | |
| FFNDMZIBVDSQFI-UHFFFAOYSA-N | |
| 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-1λâ´-chromen-1-ylium chloride | |
| 34.5-150 mm2 |
Chemical Identifiers
| 8012-95-1 | |
| 0.00 | |
| FFNDMZIBVDSQFI-UHFFFAOYSA-N | |
| * |
| MFCD00131611 | |
| MFCD00131611 | |
| 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-1λâ´-chromen-1-ylium chloride |