missing translation for 'onlineSavingsMsg'
Learn More

Mineral Oil, Heavy, USP, Spectrum™ Chemical

Catalog No. 18604608
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604608 1 L Amber Glass Bottle
18604609 4 L Amber Glass Bottle
18604610 4 x 4 L Amber Glass Bottle
18604611 20 L Amber Glass Bottle
18604612 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604608 Supplier Spectrum Chemical Mfg Cor Supplier No. MI5001LT
Get Up to $500 in Chemicals with Purchase
View all promotions

MI500, 8012-95-1

Spectrum™ Chemical Mineral Oil, Heavy, USP is a lubricant laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8012-95-1
Molecular Formula MFCD00131611
Molecular Weight (g/mol) 0.00
MDL Number MFCD00131611
InChI Key FFNDMZIBVDSQFI-UHFFFAOYSA-N
IUPAC Name 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-1λ4-chromen-1-ylium chloride
SMILES *

Specifications

Acidity To pass test
CAS Max % 1
Boiling Point >215°C / 419°F
Packaging Amber Glass Bottle
Quantity 1 L
Viscosity 34.5-150 mm2
Grade USP
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Mineral Oil, Heavy