missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Mineral Oil

Catalog No. M35161L
Encompass
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M35161L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M35161L Supplier Sigma Aldrich Supplier No. M35161L
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Mineral oils are complex mixtures of straight and branched chain paraffinic, naphthenic and aromatic hydrocarbons refined from petroleum crude oils. Mineral oil with a carbon number of 15 to 25 is categorized as light oil and 25 to 50 as heavy oil. Mineral oil is commonly used in food, cosmetics and pharmaceutical industries especially in skin care products. Mineral oil is used as overlaying agent in polymerase chain reaction (PCR) and electrophoretic techniques to avoid evaporation.Mineral oil has been used:to overlay the samples to avoid evaporation in denaturing gel PCR reaction mixin emulsion preparation for emulsion PCR

Specifications

CAS 8042-47-5
Density 0.84 g/mL at 25°C
Refractive Index n20/D 1.467
MDL Number MFCD00131611
Quantity 4 L