missing translation for 'onlineSavingsMsg'
Learn More

Milrinone 98.0+%, TCI America™

Catalog No. M16631G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16631G 1 g
M1663100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16631G Supplier TCI America Supplier No. M16631G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78415-72-2
Molecular Formula C12H9N3O
Molecular Weight (g/mol) 211.224
MDL Number MFCD00133539
InChI Key PZRHRDRVRGEVNW-UHFFFAOYSA-N
Synonym milrinone, primacor, corotrope, corotrop, milrinona, milrinonum, milrinonum latin, milrinona spanish, milrila, unii-ju9yax04c7
PubChem CID 4197
ChEBI CHEBI:50693
IUPAC Name 6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile
SMILES CC1=C(C=C(C(=O)N1)C#N)C2=CC=NC=C2

Specifications

Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 211.22
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Milrinone
RTECSNumber DW1762000
TSCA No