missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Milrinone, 97%

Chemical Identifiers

CAS 78415-72-2
Molecular Formula C12H9N3O
Molecular Weight (g/mol) 211.22
MDL Number MFCD00133539
InChI Key PZRHRDRVRGEVNW-UHFFFAOYSA-N
Synonym milrinone, primacor, corotrope, corotrop, milrinona, milrinonum, milrinonum latin, milrinona spanish, milrila, unii-ju9yax04c7
PubChem CID 4197
ChEBI CHEBI:50693
IUPAC Name 6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile
SMILES CC1=C(C=C(C(=O)N1)C#N)C2=CC=NC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC397870500
View Documents Promotion Details
Thermo Scientific™
397870500
50mg Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

78415-72-2
211.22
PZRHRDRVRGEVNW-UHFFFAOYSA-N
4197
6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile
C12H9N3O
MFCD00133539
milrinone, primacor, corotrope, corotrop, milrinona, milrinonum, milrinonum latin, milrinona spanish, milrila, unii-ju9yax04c7
CHEBI:50693
CC1=C(C=C(C(=O)N1)C#N)C2=CC=NC=C2

Specifications

78415-72-2
100.0
96% min. (HPLC)
MFCD00133539
milrinone, primacor, corotrope, corotrop, milrinona, milrinonum, milrinonum latin, milrinona spanish, milrila, unii-ju9yax04c7
CC1=C(C=C(C(=O)N1)C#N)C2=CC=NC=C2
211.22
CHEBI:50693
97%
96.0
Authentic
C12H9N3O
15, 6278
PZRHRDRVRGEVNW-UHFFFAOYSA-N
6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile
4197
211.22
Milrinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Fatal if inhaled.
Toxic in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 278-903-6

SDS
Documents

Documents

RUO – Research Use Only