missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MilliSentials™ Lab Labeling System

Catalog No. 09735143
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
09735143 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Improper use of the instrument or the battery (short-circuit, mechanical damage, overheating etc.) may cause the explosion of the battery. Only use manufacturer’s lithium ion battery.
Catalog No. 09735143 Supplier MilliporeSigma™ Supplier No. MISELLABSN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Improper use of the instrument or the battery (short-circuit, mechanical damage, overheating etc.) may cause the explosion of the battery. Only use manufacturer’s lithium ion battery.

The MilliSentials™ Lab Labeling System provides a complete laboratory labeling solution with laboratory grade labels, a compact WiFi capable printer, and custom-designed laboratory labeling software.

  • Laboratory labels are capable of withstanding temperatures from -196°C (liquid nitrogen) to 100°C (boiling water) and provide resistance to harsh chemical solvents (including ethanol, DMSO, xylene)
  • Labels have been designed with perforations, allowing one size to fit across a wide range of laboratory vessels including microcentrifuge tubes, cryovials, petri dishes, cell culture plates, and reagent bottles
  • Custom-built software features preconstructed label templates for laboratory vessels, allows labels to be saved for quick repeat printing
  • Features a spreadsheet upload function for large batch printing
  • System offers a complete labeling system specifically designed for the laboratory
  • Connection via WiFi, Bluetooth™ 4.1, or USB
  • Ribbon Capacity: 6400 labels printed from 1 roll of ribbon

Specifications

Product Type Label Printer
Color White
Display Type Digital
Length (English) 10.5 in.
Width (English) 8 in.
Width (Metric) 202 mm
Dimensions (L x W) 10.5 x 8 in.
Adhesive Type Hot-melt
Sterility Non-sterile
Color Capability Grayscale
Includes MilliSentials Lab Printer, 1 Roll Lab Adaptable Labels, 1 Roll Lab Printer Ribbon, Lab Labeling System Software, USB Key
Languages English
Printable Sides One
Print Speed 6 in./sec.
Resolution 300 dpi
Shape Rectangle with Square Corners
Weight (Metric) 2.5 kg
Warranty 3 Years
Power Supply Details Auto-detectable (PFC Compliant)
Voltage 100/240 V
Show More Show Less

Unless otherwise stated, products are intended for research use only and are not to be used for any other purpose, including but not limited to unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses, or any type of consumption or application to humans or animals.