missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Spring Clamp for Vacuum Filtration
Description
Millipore Glass Vacuum Filtration Accessory Product
Spring Clamp, 47 mm, Aluminum
These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
This item is an accessory part for the Millipore 47 mm Classic Glass Filter Holder Kits, Millipore 47 mm All-Glass Filter Holder Kits and Millipore MilliSolve Kit.
Specifications
Specifications
| Type | Laboratory Filter Holders or Cyclones |
| Material | Anodized Aluminum |
| For Use With (Equipment) | Millipore Classic Glass Filter Holder Kits, All-Glass Filter Holder Kits and Millipore MilliSolve Ki |
| For Use With (Application) | Vacuum filtration |
| Diameter (Metric) | 47 mm |
| Color | Clear |
| Diameter (Metric) Inside | 47 mm |