Learn More
MilliporeSigma Millicell™ Disposable Hemocytometer
Catalog No. MDH4N150PK
Change view
Click to view available options
Includes:
2 Chambers/Slide
4 Chambers/Slide
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
MDH4N150PK 4 Chambers/Slide
MDH2N150PK 2 Chambers/Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MDH4N150PK Supplier MilliporeSigma Supplier No. MDH4N150PK
Only null left
Individually wrapped Neubauer Improved hemocytometer for counting cells.
The Millicell™ disposable hemocytometer is used to manually count cells. It consists of a surface-patterned two or four enclosed chambers with ports for sample injection. Count cells in the same way as you do with the Neubauer Improved counting chamber.
The Neubauer Improved counting chamber consists of 9 large squares, each measuring 1 x 1 mm. Each square has a total volume of 0.1 mm3. The cells are distributed over 3 x 3 large squares, each with 1 mm edge length and with a surface area of 1 mm2.
With the Millicell™ disposable hemocytometer, you inject 10 μL of sample, stained or unstained, into the desired chamber.
Specifications
Product Type Hemacytometer
Grid Type Neubauer Improved
For Use With (Application) Cell counting
Includes 4 Chambers/Slide