Learn More
MilliporeSigma Millicell™ Digital Cell Imager
Digital Cell Imager allows you to assess your cell cultures with ease.
Supplier:  MilliporeSigma MDCI10000
Catalog No. MDCI10000
Only null left
Explore available promotions
Description
Description
  • The Millicell™ DCI instrument enables more efficient execution of the repetitive daily techniques associated with cell passaging.
  • Quickly and objectively assess confluency, morphology, and growth trends for more consistent cell cultures.
  • Remove subjectivity by using accurate confluency measurement.
  • WiFi compatible with the included EDIMAX external USB and Ethernet compatible.
Available software subscriptions:
  • Trial - Catalog # 09-804-165
  • 1 year - Catalog # 09-804-167
  • 5 year - Catalog # 09-804-166
Specifications
Specifications
Cell Imaging System
Brightfield
Product Suggestions
Product Suggestions