Learn More
MilliporeSigma Milli-Q™ SQ Cleaning Tool
Catalog No. ZSQ240CTT0
Change view
Click to view available options
Type: Cleanign Tool
Catalog No.
ZSQ240CTT0 Type: Cleanign Tool
1 options
Catalog No. ZSQ240CTT0 Supplier MilliporeSigma Supplier No. ZSQ240CTT0
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Sanitization of the Milli-Q™ SQ Switch Tank and Milli-Q™ SQ/SR production station
  • The Milli-Q™ SQ Cleaning Tool is a specially dedicated tool for the sanitization of the Milli-Q™ SQ/SR production station and its Milli-Q™ SQ Switch Tank.
  • It uses chlorine tablets, such ROProtect C or EfferSan (not included) for the sanitization process.
  • There is a specific procedure to be followed for the usage of this tool.
Specifications
Type Cleaning Tool